siRNA Details
Gene Symbol: SYT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146261.2 | NP_001139733.1 | 4 | 292 | ||
NM_001146262.2 | NP_001139734.1 | 3 | 171 | |||
NM_001146264.2 | NP_001139736.1 | 4 | 292 | |||
NM_001256006.1 | NP_001242935.1 | 4 | 376 | |||
NM_153262.3 | NP_694994.2 | 3 | 171 | |||
NR_027459.2 | 4 | 376 | ||||
XM_017000932.1 | XP_016856421.1 | 4 | 366 | |||
XM_017000933.1 | XP_016856422.1 | 4 | 229 | |||
XM_017000934.1 | XP_016856423.1 | 1 | 25 | |||
XM_017000935.1 | XP_016856424.1 | 3 | 180 | |||
GenBank mRNA | AB102948.1 | - | 2 | 80 | ||
AJ617623.1 | - | 3 | 114 | |||
AJ617624.1 | - | 3 | 257 | |||
AJ617625.1 | - | 4 | 290 | |||
AK091517.1 | - | 3 | 94 | |||
BC130323.1 | - | 4 | 225 | 40146354 | ||
BC144154.1 | - | 4 | 225 | |||
BC144155.1 | - | 3 | 104 | |||
BC144157.1 | - | 4 | 225 |

More Information
Other Species Targeted

Gene Ontology Categories:
