siRNA Details
Gene Symbol: DEFB119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153289.3 | NP_695021.2 | 2 | 359 | ||
NR_073151.1 | 4 | 436 | ||||
NR_073152.1 | 4 | 313 | ||||
NR_073153.1 | 3 | 399 | ||||
NR_126440.1 | 2 | 271 | ||||
GenBank mRNA | AF479698.1 | - | 2 | 193 | ||
AL732301.1 | - | 4 | 356 | |||
AY122472.1 | - | 2 | 313 | |||
AY358860.1 | - | 2 | 285 | |||
AY501001.1 | - | 3 | 279 | |||
BC062212.1 | - | 2 | 304 | 6621528 | ||
BI460707.1 | - | 3 | 344 | |||
BU561156.1 | - | 4 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
