siRNA Details
Gene Symbol: ABCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000350.2 | NP_000341.2 | 3 | 374 | ||
GenBank mRNA | AB210040.1 | - | 3 | 360 | ||
AF000148.1 | - | 3 | 357 | |||
AF001945.1 | - | 3 | 351 | |||
AK295818.1 | - | 3 | 350 | |||
AL712362.1 | - | 3 | 374 | |||
U88667.1 | - | 3 | 351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
