siRNA Details
Gene Symbol: ZDHHC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286638.1 | NP_001273567.1 | 11 | 4092 | ||
NM_153251.3 | NP_694983.2 | 12 | 4115 | |||
NR_104486.1 | 15 | 4360 | ||||
NR_104487.1 | 13 | 4164 | ||||
XM_006719795.1 | XP_006719858.1 | 13 | 4136 | |||
XM_006719796.1 | XP_006719859.1 | 13 | 4133 | |||
XM_006719797.1 | XP_006719860.1 | 12 | 4058 | |||
XM_011535030.2 | XP_011533332.1 | 7 | 3615 | |||
XM_017020525.1 | XP_016876014.1 | 11 | 3829 | |||
XM_017020526.1 | XP_016876015.1 | 14 | 3902 | |||
XM_017020527.1 | XP_016876016.1 | 12 | 4034 | |||
GenBank mRNA | AK090979.1 | - | 1 | 964 | ||
AK098818.1 | - | 2 | 928 | |||
AL833626.1 | - | 4 | 2124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
