siRNA Details
Gene Symbol: SNX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153337.2 | NP_699168.1 | 4 | 477 | ||
GenBank mRNA | AK055837.1 | - | 4 | 442 | ||
AK292705.1 | - | 4 | 442 | |||
BC027944.1 | - | 4 | 426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
