siRNA Details
Gene Symbol: TMEM92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168215.1 | NP_001161687.1 | 6 | 1260 | ||
NM_153229.2 | NP_694961.2 | 5 | 1225 | |||
XM_006721730.3 | XP_006721793.1 | 5 | 1183 | |||
GenBank mRNA | AK090637.1 | - | 5 | 1180 |
