siRNA Details
Gene Symbol: LOC439933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122079.1 | 5 | 1023 | |||
NR_122080.1 | 3 | 898 | ||||
GenBank mRNA | BC030111.1 | - | 3 | 904 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
