siRNA Details
Gene Symbol: ERI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153332.3 | NP_699163.2 | 7 | 4329 | ||
XM_005272401.2 | XP_005272458.1 | 7 | 4184 | |||
XM_011543849.1 | XP_011542151.1 | 7 | 1506 | |||
XM_011543850.2 | XP_011542152.1 | 7 | 4368 | |||
XM_011543851.1 | XP_011542153.1 | 8 | 4383 | |||
XM_017013949.1 | XP_016869438.1 | 11 | 5276 | |||
XM_017013950.1 | XP_016869439.1 | 7 | 5013 | |||
XM_017013951.1 | XP_016869440.1 | 8 | 4735 | |||
XM_017013952.1 | XP_016869441.1 | 8 | 5056 | |||
XM_017013953.1 | XP_016869442.1 | 8 | 4306 | |||
XM_017013954.1 | XP_016869443.1 | 8 | 5160 | |||
GenBank mRNA | AK226180.1 | - | 7 | 4197 | ||
AL137679.1 | - | 8 | 1823 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
