siRNA Details
Gene Symbol: IFNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000605.3 | NP_000596.2 | 1 | 292 | ||
GenBank mRNA | AY255838.1 | - | 1 | 158 | ||
BC074936.2 | - | 1 | 268 | 30915269 | ||
BC074937.2 | - | 1 | 268 | 30915492 | ||
BC104163.1 | - | 1 | 269 | 40029595 | ||
BC104164.1 | - | 1 | 269 | 40029596 | ||
CR541921.1 | - | 1 | 224 | |||
JN591568.1 | - | 1 | 224 | |||
JN591569.1 | - | 1 | 224 | |||
JN591570.1 | - | 1 | 224 | |||
JN848522.1 | - | 1 | 224 | |||
M54886.1 | - | 1 | 284 | |||
V00544.1 | - | 1 | 284 | |||
V00548.1 | - | 1 | 206 | |||
V00549.1 | - | 1 | 284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
