siRNA Details
Gene Symbol: RARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000966.5 | NP_000957.1 | 6 | 1010 | ||
NM_001042728.2 | NP_001036193.1 | 4 | 798 | |||
NM_001243730.1 | NP_001230659.1 | 5 | 684 | |||
NM_001243731.1 | NP_001230660.1 | 4 | 535 | |||
NM_001243732.1 | NP_001230661.1 | 3 | 649 | |||
GenBank mRNA | AB307689.1 | - | 4 | 518 | ||
AK290588.1 | - | 6 | 1006 | |||
AK297023.1 | - | 5 | 602 | |||
AK297110.1 | - | 4 | 511 | |||
AK297277.1 | - | 3 | 649 | |||
AK301057.1 | - | 4 | 793 | |||
AK316259.1 | - | 4 | 522 | |||
BC093727.1 | - | 4 | 542 | 7939572 | ||
BC093729.1 | - | 4 | 542 | 7939574 | ||
HQ692828.1 | - | 4 | 518 | |||
HQ692829.1 | - | 4 | 485 | |||
L12060.1 | - | 5 | 626 | |||
M24857.1 | - | 6 | 932 | |||
M38258.1 | - | 4 | 664 | |||
M57707.1 | - | 5 | 717 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
