siRNA Details
Gene Symbol: BAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182482.2 | NP_872288.2 | 8 | 1674 | ||
GenBank mRNA | AF218570.1 | - | 8 | 1674 |
Gene Symbol: BAGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182481.1 | NP_872287.1 | 8 | 1674 | ||
GenBank mRNA | AF339514.1 | - | 8 | 1674 |
Gene Symbol: BAGE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182484.1 | NP_872290.1 | 7 | 1623 | ||
GenBank mRNA | AF339516.1 | - | 7 | 1623 |
Gene Symbol: LOC101927050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136329.1 | 2 | 129 | |||
GenBank mRNA | DB034577.1 | - | 2 | 129 |
Gene Symbol: LOC101927429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136328.1 | 2 | 129 | |||
GenBank mRNA | AK310441.1 | - | 2 | 132 |
Gene Symbol: LOC105378260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028520.1 | XP_016884009.1 | 5 | 1125 | ||
XM_017028521.1 | XP_016884010.1 | 5 | 1125 |
Gene Symbol: LOC107983967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003062.1 | XP_016858551.1 | 2 | 152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
