siRNA Details
Gene Symbol: HS3ST5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153612.3 | NP_705840.2 | 2 | 2193 | ||
XM_006715379.2 | XP_006715442.1 | 5 | 2681 | |||
XM_011535588.2 | XP_011533890.1 | 3 | 3397 | |||
XM_017010470.1 | XP_016865959.1 | 5 | 3255 | |||
XM_017010471.1 | XP_016865960.1 | 5 | 4214 | |||
XM_017010472.1 | XP_016865961.1 | 4 | 3851 | |||
XM_017010473.1 | XP_016865962.1 | 5 | 3251 | |||
XM_017010474.1 | XP_016865963.1 | 5 | 2689 | |||
GenBank mRNA | AB074164.1 | - | 1 | 72 | ||
AK091074.1 | - | 1 | 1709 | |||
AK126930.1 | - | 2 | 2851 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
