siRNA Details
Gene Symbol: CPNE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153635.2 | NP_705899.2 | 21 | 1842 | ||
XM_011533386.2 | XP_011531688.1 | 20 | 2018 | |||
XM_017005747.1 | XP_016861236.1 | 20 | 1602 | |||
GenBank mRNA | BC130375.1 | - | 21 | 1713 | 40146406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
