siRNA Details
Gene Symbol: LOC146880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027487.2 | 3 | 1734 | |||
GenBank mRNA | AK097700.1 | - | 2 | 984 | ||
BC032382.1 | - | 3 | 1734 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
