siRNA Details
Gene Symbol: CCDC150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006712439.3 | XP_006712502.1 | 1 | 449 | ||
GenBank mRNA | AK125768.1 | - | 1 | 449 | ||
AL834537.1 | - | 1 | 344 | |||
BC062658.1 | - | 1 | 405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
