siRNA Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198622.1 | NP_001185551.1 | 3 | 1149 | ||
NM_001198623.1 | NP_001185552.1 | 3 | 1146 | |||
NM_001198624.1 | NP_001185553.1 | 4 | 454 | |||
NM_003808.3 | NP_003799.1 | 4 | 1230 | |||
NM_172087.2 | NP_742084.1 | 3 | 1182 | |||
NM_172088.2 | NP_742085.1 | 4 | 1230 | |||
NR_073490.2 | 2 | 1103 | ||||
GenBank mRNA | AF046888.1 | - | 4 | 763 | ||
AF114011.1 | - | 4 | 763 | |||
AF114012.1 | - | 3 | 631 | |||
AF114013.1 | - | 4 | 735 | |||
AF136294.1 | - | 4 | 482 | |||
AF184972.1 | - | 4 | 607 | |||
AK090698.1 | - | 3 | 998 | |||
AK225556.1 | - | 4 | 774 | |||
AK301221.1 | - | 3 | 679 | |||
AK314518.1 | - | 4 | 770 | |||
AY081050.1 | - | 4 | 754 | |||
AY358880.1 | - | 4 | 595 | |||
AY377858.1 | - | 3 | 170 | |||
BC008042.2 | - | 4 | 585 | 3837834 | ||
BF793371.1 | - | 452 | ||||
BT019561.1 | - | 4 | 482 | |||
BT019562.1 | - | 4 | 482 | |||
CR542040.1 | - | 4 | 482 | |||
CR542073.1 | - | 4 | 482 | |||
DQ149579.1 | - | 3 | 431 | |||
EF211088.1 | - | 2 | 385 |
Gene Symbol: TNFSF12-TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172089.3 | NP_742086.1 | 9 | 818 | ||
GenBank mRNA | AY081051.1 | - | 9 | 777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
