siRNA Details
Gene Symbol: ADGRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001704.2 | NP_001695.1 | 3 | 684 | ||
XM_005248752.2 | XP_005248809.1 | 2 | 970 | |||
XM_006715534.3 | XP_006715597.1 | 3 | 1106 | |||
XM_011536010.2 | XP_011534312.1 | 3 | 837 | |||
XM_011536011.2 | XP_011534313.1 | 2 | 970 | |||
XM_011536012.2 | XP_011534314.1 | 3 | 1106 | |||
XM_017011140.1 | XP_016866629.1 | 2 | 934 | |||
XM_017011142.1 | XP_016866631.1 | 3 | 1106 | |||
XM_017011143.1 | XP_016866632.1 | 3 | 1106 | |||
XM_017011144.1 | XP_016866633.1 | 3 | 1106 | |||
GenBank mRNA | AB005299.1 | - | 2 | 269 | ||
AB011122.2 | - | 3 | 473 | |||
AK289696.1 | - | 2 | 285 | |||
BC111720.1 | - | 1 | 263 | 40027602 | ||
DB446627.2 | - | 2 | 513 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175642, AY168406, BC099951.
Set Membership: |
![]() |
Gene Ontology Categories:
