siRNA Details
Gene Symbol: TRRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244580.1 | NP_001231509.1 | 7 | 706 | ||
NM_003496.3 | NP_003487.1 | 7 | 706 | |||
GenBank mRNA | AF076974.1 | - | 7 | 706 | ||
AF110377.1 | - | 7 | 626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
