siRNA Details
Gene Symbol: C10orf107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173554.2 | NP_775825.1 | 3 | 403 | ||
XM_005269598.3 | XP_005269655.1 | 4 | 393 | |||
XM_005269599.4 | XP_005269656.1 | 4 | 501 | |||
XM_005269600.4 | XP_005269657.1 | 3 | 304 | |||
XM_006717690.3 | XP_006717753.1 | 4 | 375 | |||
XM_006717692.3 | XP_006717755.1 | 4 | 375 | |||
GenBank mRNA | BC041932.1 | - | 3 | 418 | 5300631 |
