siRNA Details
Gene Symbol: ANKRD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173595.3 | NP_775866.2 | 3, 4 | 288 | ||
XM_011538197.2 | XP_011536499.1 | 2, 3 | 390 | |||
XM_017019183.1 | XP_016874672.1 | 2, 3 | 471 | |||
XM_017019184.1 | XP_016874673.1 | 3, 4 | 320 | |||
GenBank mRNA | AB190990.1 | - | 3, 4 | 305 | ||
AK125379.1 | - | 3, 4 | 305 | |||
DR002390.1 | - | 3, 4 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
