siRNA Details
Gene Symbol: HNF4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000457.4 | NP_000448.3 | 10 | 2103 | ||
NM_001030003.2 | NP_001025174.1 | 10 | 1894 | |||
NM_001258355.1 | NP_001245284.1 | 11 | 2194 | |||
NM_001287182.1 | NP_001274111.1 | 11 | 2116 | |||
NM_001287183.1 | NP_001274112.1 | 11 | 2146 | |||
NM_175914.4 | NP_787110.2 | 10 | 1924 | |||
NM_178849.2 | NP_849180.1 | 10 | 2073 | |||
XM_005260407.3 | XP_005260464.1 | 11 | 2223 | |||
GenBank mRNA | X87870.1 | - | 10 | 2016 | ||
Z49825.1 | - | 11 | 2173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
