siRNA Details
Gene Symbol: ENDOV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164637.2 | NP_001158109.1 | 2, 3 | 264 | ||
NM_001164638.1 | NP_001158110.1 | 2, 3 | 264 | |||
XM_005257246.3 | XP_005257303.1 | 2, 3 | 266 | |||
XM_011524660.1 | XP_011522962.1 | 2, 3 | 264 | |||
XM_011524661.2 | XP_011522963.1 | 2, 3 | 266 | |||
XM_011524672.1 | XP_011522974.1 | 2, 3 | 247 | |||
XM_011524674.1 | XP_011522976.1 | 2, 3 | 247 | |||
XM_011524678.2 | XP_011522980.1 | 1, 2 | 362 | |||
XM_017024511.1 | XP_016880000.1 | 2, 3 | 266 | |||
XM_017024514.1 | XP_016880003.1 | 2, 3 | 247 | |||
XM_017024518.1 | XP_016880007.1 | 2, 3 | 247 | |||
GenBank mRNA | AK092539.1 | - | 2, 3 | 264 | ||
AK096802.1 | - | 1, 2 | 132 | |||
BC064545.1 | - | 2, 3 | 247 | 5736957 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
