siRNA Details
Gene Symbol: ZNF776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173632.3 | NP_775903.3 | 3 | 1644 | ||
GenBank mRNA | AK095607.1 | - | 1 | 468 | ||
AK127764.1 | - | 2 | 1290 | |||
AK299680.1 | - | 3 | 1644 | |||
BC136752.1 | - | 2 | 1286 | 9020742 | ||
BC136753.1 | - | 2 | 1286 | 9020743 | ||
BX957217.1 | - | 3 | 1683 | |||
CR749390.1 | - | 3 | 1683 | |||
CR936802.1 | - | 3 | 1598 |
