siRNA Details
Gene Symbol: CCDC150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080539.1 | NP_001074008.1 | 2 | 247 | ||
XM_006712438.3 | XP_006712501.1 | 2 | 247 | |||
XM_017003861.1 | XP_016859350.1 | 2 | 247 | |||
XM_017003862.1 | XP_016859351.1 | 2 | 247 | |||
XM_017003863.1 | XP_016859352.1 | 2 | 247 | |||
XM_017003864.1 | XP_016859353.1 | 2 | 247 | |||
GenBank mRNA | AK096979.1 | - | 2 | 247 | ||
AK299214.1 | - | 2 | 164 | |||
AK302685.1 | - | 2 | 325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
