siRNA Details
Gene Symbol: ZNF619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145082.2 | NP_001138554.1 | 5, 6 | 730 | ||
NM_001145083.1 | NP_001138555.1 | 3, 4 | 427 | |||
NM_001145093.2 | NP_001138565.1 | 4, 5 | 610 | |||
NR_026746.2 | 5, 6 | 760 | ||||
XM_006713112.2 | XP_006713175.1 | 4, 5 | 578 | |||
XM_006713113.3 | XP_006713176.1 | 3, 4 | 434 | |||
XM_011533607.1 | XP_011531909.1 | 5, 6 | 698 | |||
XM_011533608.1 | XP_011531910.1 | 5, 6 | 652 | |||
XM_017006225.1 | XP_016861714.1 | 4, 5 | 576 | |||
GenBank mRNA | AK075245.1 | - | 5, 6 | 627 | ||
AK304139.1 | - | 3, 4 | 427 | |||
AK304741.1 | - | 5, 6 | 689 | |||
BC117169.1 | - | 3, 4 | 362 | 40125720 | ||
BC143304.1 | - | 2, 3 | 291 | |||
BC143306.1 | - | 3, 4 | 362 | 9051811 | ||
BC143331.1 | - | 3, 4 | 441 |
