siRNA Details
Gene Symbol: SSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_198455.2 | NP_940857.2 | 92 | 14012 | ||
GenBank mRNA | AK093431.1 | - | 3 | 293 | ||
AK123170.1 | - | 8 | 1025 | |||
AK124542.1 | - | 2 | 894 | |||
AY927596.1 | - | LIT3589 AY927596 |
2 | 77 | ||
AY927597.1 | - | LIT3590 AY927597 |
3 | 202 | ||
AY927598.1 | - | 7 | 819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
