siRNA Details
Gene Symbol: FAM9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205849.3 | NP_995321.1 | 6 | 745 | ||
XM_011545464.2 | XP_011543766.1 | 13 | 1318 | |||
GenBank mRNA | AF494344.1 | - | 6 | 744 | ||
AK097501.1 | - | 5 | 689 | |||
AK128470.1 | - | 6 | 3106 | |||
BC120955.1 | - | 5 | 444 | 40115245 | ||
BC120956.1 | - | 5 | 444 | 40115246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
