siRNA Details
Gene Symbol: FAM133A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171109.1 | NP_001164580.1 | 4 | 694 | ||
NM_001171110.1 | NP_001164581.1 | 5 | 729 | |||
NM_001171111.1 | NP_001164582.1 | 3 | 605 | |||
NM_173698.2 | NP_775969.1 | 4 | 674 | |||
XM_006724641.3 | XP_006724704.1 | 3 | 706 | |||
XM_011530925.2 | XP_011529227.1 | 4 | 647 | |||
XM_011530926.2 | XP_011529228.1 | 4 | 485 | |||
XM_011530927.2 | XP_011529229.1 | 4 | 524 | |||
XM_011530928.1 | XP_011529230.1 | 4 | 458 | |||
XM_011530929.1 | XP_011529231.1 | 5 | 493 | |||
XM_011530931.2 | XP_011529233.1 | 3 | 660 | |||
XM_017029454.1 | XP_016884943.1 | 5 | 3333 | |||
XM_017029455.1 | XP_016884944.1 | 6 | 666 | |||
XM_017029456.1 | XP_016884945.1 | 4 | 601 | |||
GenBank mRNA | AK094978.1 | - | 4 | 481 | ||
BC113385.1 | - | 1 | 191 | 8322437 | ||
BC113387.1 | - | 1 | 191 | 8322439 | ||
DA188870.1 | - | 5 | 539 | |||
DA297338.1 | - | 4 | 481 | |||
DA414074.1 | - | 4 | 504 | |||
DB084469.1 | - | 3 | 416 |
