siRNA Details
Gene Symbol: ARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282380.1 | NP_001269309.1 | 6 | 2073 | ||
NM_182616.3 | NP_872422.1 | 6 | 1684 | |||
GenBank mRNA | AK093274.1 | - | 6 | 2073 | ||
BC053602.1 | - | 6 | 1621 | 5770387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
