siRNA Details
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 7, 8 | 681 | ||
XM_011545742.2 | XP_011544044.1 | 9, 10 | 828 | |||
XM_017022947.1 | XP_016878436.1 | 10, 11 | 901 | |||
GenBank mRNA | AK098260.1 | - | 6, 7 | 774 | ||
AK307544.1 | - | 7, 8 | 677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
