siRNA Details
Gene Symbol: HMGCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY429543.1 | - | 4 | 603 | ||
M11058.1 | - | 20 | 2823 | |||
M62633.1 | - | 1 | 109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
