siRNA Details
Gene Symbol: ZNF660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173658.2 | NP_775929.2 | 3 | 473 | ||
XM_005265090.4 | XP_005265147.1 | 2 | 571 | |||
XM_006713117.3 | XP_006713180.1 | 3 | 572 | |||
XM_011533622.2 | XP_011531924.1 | 3 | 776 | |||
GenBank mRNA | AK094189.1 | - | 3 | 473 | ||
BC075094.2 | - | 1 | 164 | 30915605 | ||
BC075095.2 | - | 1 | 164 | 30915393 |
