siRNA Details
Gene Symbol: C8orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013842.2 | NP_001013864.1 | 6 | 1031 | ||
NM_173686.2 | NP_775957.2 | 5, 6 | 1007 | |||
GenBank mRNA | AK092034.1 | - | 4, 5 | 815 | ||
AK295405.1 | - | 5, 6 | 1007 | |||
BC012750.2 | - | 5 | 822 | 3618901 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
