siRNA Details
Gene Symbol: PROX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080408.2 | NP_001073877.2 | 3 | 1210 | ||
NM_001243007.1 | NP_001229936.1 | 4 | 1891 | |||
GenBank mRNA | AK094068.1 | - | 3 | 894 | ||
BC105720.1 | - | 4 | 1890 | |||
BC105927.1 | - | 4 | 1890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
