siRNA Details
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 14 | 1416 | ||
XM_011545742.2 | XP_011544044.1 | 16 | 1563 | |||
XM_011545744.2 | XP_011544046.1 | 1 | 192 | |||
XM_011545746.2 | XP_011544048.1 | 1 | 192 | |||
XM_017022947.1 | XP_016878436.1 | 17 | 1636 | |||
GenBank mRNA | AK098260.1 | - | 12 | 1373 | ||
AK128606.1 | - | 3 | 284 | |||
AK307544.1 | - | 14 | 1412 | |||
BC109294.1 | - | 2 | 144 | |||
BC109295.1 | - | 2 | 144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
