siRNA Details
Gene Symbol: CCDC150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080539.1 | NP_001074008.1 | 3 | 518 | ||
XM_006712438.3 | XP_006712501.1 | 3 | 518 | |||
XM_011510985.2 | XP_011509287.1 | 2 | 310 | |||
XM_011510988.2 | XP_011509290.1 | 2 | 289 | |||
XM_017003861.1 | XP_016859350.1 | 3 | 518 | |||
XM_017003862.1 | XP_016859351.1 | 3 | 518 | |||
XM_017003863.1 | XP_016859352.1 | 3 | 518 | |||
XM_017003864.1 | XP_016859353.1 | 3 | 518 | |||
XM_017003865.1 | XP_016859354.1 | 2 | 289 | |||
GenBank mRNA | AK096979.1 | - | 3 | 518 | ||
AK303236.1 | - | 2 | 284 | |||
AK309349.1 | - | 2 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
