siRNA Details
Gene Symbol: CCDC141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173648.3 | NP_775919.3 | 24 | 5735 | ||
XM_011510991.2 | XP_011509293.1 | 25 | 6027 | |||
XM_017003871.1 | XP_016859360.1 | 13 | 3947 | |||
GenBank mRNA | AK096821.1 | - | 3 | 2269 | ||
BX649042.1 | - | 13 | 3337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
