siRNA Details
Gene Symbol: RGMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543347.2 | XP_011541649.2 | 3 | 862 | ||
GenBank mRNA | AK074887.1 | - | 4 | 1425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
