siRNA Details
Gene Symbol: FAM9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205849.3 | NP_995321.1 | 9 | 2023 | ||
GenBank mRNA | AK097501.1 | - | 8 | 1967 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
