siRNA Details
Gene Symbol: DPY19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173812.4 | NP_776173.3 | 1 | 285 | ||
XM_017019188.1 | XP_016874677.1 | 1 | 320 | |||
XM_017019189.1 | XP_016874678.1 | 1 | 320 | |||
XM_017019190.1 | XP_016874679.1 | 1 | 320 | |||
XM_017019191.1 | XP_016874680.1 | 1 | 320 | |||
XM_017019192.1 | XP_016874681.1 | 1 | 320 | |||
XM_017019196.1 | XP_016874685.1 | 1 | 320 | |||
XM_017019197.1 | XP_016874686.1 | 1 | 253 | |||
XM_017019198.1 | XP_016874687.1 | 1 | 320 | |||
XM_017019199.1 | XP_016874688.1 | 1 | 253 | |||
XM_017019200.1 | XP_016874689.1 | 1 | 320 | |||
XM_017019201.1 | XP_016874690.1 | 1 | 220 | |||
XM_017019203.1 | XP_016874692.1 | 1 | 115 | |||
XM_017019204.1 | XP_016874693.1 | 1 | 253 | |||
XM_017019205.1 | XP_016874694.1 | 1 | 320 | |||
GenBank mRNA | AK057511.1 | - | 1 | 256 | ||
AK310102.1 | - | 1 | 255 | |||
BC125215.1 | - | 1 | 175 | |||
BC125216.1 | - | 1 | 175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
