siRNA Details
Gene Symbol: TCAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282913.1 | NP_001269842.1 | 4 | 444 | ||
NM_001282914.1 | NP_001269843.1 | 4 | 626 | |||
NM_001282915.1 | NP_001269844.1 | 4 | 444 | |||
NM_173826.3 | NP_776187.2 | 4 | 626 | |||
XM_017006251.1 | XP_016861740.1 | 5 | 1654 | |||
GenBank mRNA | AK292756.1 | - | 4 | 444 | ||
AL832473.1 | - | 4 | 303 | |||
AL833118.1 | - | 2 | 116 | |||
BC047231.1 | - | 4 | 321 | 4817904 | ||
BC101058.2 | - | 4 | 303 | |||
BC101059.2 | - | 4 | 303 | 40008143 | ||
BC101060.2 | - | 4 | 303 | 40008144 | ||
BC101061.2 | - | 4 | 303 | 40008152 | ||
CN372173.1 | - | 4 | 399 | |||
DC413255.1 | - | 4 | 444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
