siRNA Details
Gene Symbol: CCDC85C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144995.1 | NP_001138467.1 | 6 | 2218 | ||
XM_011536705.2 | XP_011535007.1 | 7 | 1901 | |||
GenBank mRNA | AK093969.1 | - | 1 | 361 | ||
AK123840.1 | - | 5 | 1106 | |||
BC110042.1 | - | 1 | 484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
