siRNA Details
Gene Symbol: POTED | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174981.3 | NP_778146.2 | 1 | 525 | ||
XM_006723997.3 | XP_006724060.1 | 1 | 621 | |||
XM_011529550.2 | XP_011527852.1 | 1 | 621 |
Gene Symbol: LOC100288966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257362.1 | NP_001244291.1 | 1 | 525 | ||
XM_006709930.3 | XP_006709993.1 | 1 | 621 | |||
XM_011545655.2 | XP_011543957.1 | 1 | 621 | |||
XM_011545656.2 | XP_011543958.1 | 1 | 621 | |||
GenBank mRNA | AY172978.1 | - | 1 | 525 |
Gene Symbol: LOC102723502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724900.3 | XP_006724963.1 | 1 | 620 | ||
XM_006724901.3 | XP_006724964.1 | 1 | 620 | |||
XM_011546199.2 | XP_011544501.1 | 1 | 620 | |||
XM_011546200.2 | XP_011544502.1 | 1 | 620 |
Gene Symbol: POTEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277304.1 | NP_001264233.1 | 1 | 395 | ||
NR_102391.1 | 1 | 395 | ||||
XM_006720355.3 | XP_006720418.1 | 1 | 509 | |||
XM_011543794.2 | XP_011542096.1 | 1 | 509 | |||
XM_017021858.1 | XP_016877347.1 | 1 | 509 |
Gene Symbol: POTEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277303.1 | NP_001264232.1 | 1 | 395 | ||
NR_102390.1 | 1 | 395 | ||||
XM_006720354.3 | XP_006720417.1 | 1 | 509 | |||
XM_011543793.2 | XP_011542095.1 | 1 | 509 | |||
XM_017021853.1 | XP_016877342.1 | 1 | 509 | |||
GenBank mRNA | BC101007.1 | - | 1 | 395 | 40006489 |
Gene Symbol: POTEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207355.3 | NP_997238.2 | 1 | 844 | ||
NR_110752.1 | 1 | 844 | ||||
XM_006720358.2 | XP_006720421.1 | 1 | 891 | |||
XM_011543795.2 | XP_011542097.1 | 1 | 891 | |||
XM_011543796.2 | XP_011542098.1 | 1 | 891 | |||
XM_011543797.2 | XP_011542099.1 | 1 | 891 | |||
GenBank mRNA | AY465170.1 | - | 2 | 414 | ||
AY465173.1 | - | 1 | 521 | |||
BC066981.1 | - | 1 | 844 | 4824630 | ||
BC101005.1 | - | 1 | 395 | 40006483 | ||
BC101006.1 | - | 1 | 506 | 40006485 | ||
BC127123.1 | - | 1 | 506 | 40006490 |
Gene Symbol: POTEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137671.1 | NP_001131143.1 | 1 | 927 | ||
NR_134878.1 | 1 | 927 | ||||
GenBank mRNA | AK307658.1 | - | 1 | 927 | ||
BC140940.1 | - | 1 | 820 | 9056752 | ||
BX649118.1 | - | 1 | 913 |
Gene Symbol: POTEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083538.1 | NP_001077007.1 | 1 | 525 | ||
XM_011511216.2 | XP_011509518.1 | 1 | 603 | |||
XM_017004159.1 | XP_016859648.1 | 1 | 604 | |||
XM_017004161.1 | XP_016859650.1 | 3 | 962 | |||
GenBank mRNA | AY462868.1 | - | 1 | 525 | ||
AY462871.1 | - | 1 | 525 | |||
AY462872.1 | - | 1 | 525 | |||
AY462873.1 | - | 1 | 525 |
Gene Symbol: POTEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099771.2 | NP_001093241.1 | 3 | 873 | ||
XM_017004833.1 | XP_016860322.1 | 1 | 618 | |||
GenBank mRNA | EF523384.1 | - | 3 | 875 |
Gene Symbol: POTEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005356.2 | NP_001005356.1 | 1 | 525 | ||
NR_027480.1 | 1 | 525 | ||||
GenBank mRNA | AY462874.1 | - | 1 | 525 | ||
AY465171.1 | - | 2 | 525 | |||
AY465172.1 | - | 2 | 525 | |||
BC127623.1 | - | 1 | 505 |
Gene Symbol: POTEH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136213.1 | NP_001129685.1 | 1 | 636 | ||
GenBank mRNA | AY466021.1 | - | 1 | 636 |
Gene Symbol: POTEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145442.1 | NP_001138914.1 | 1 | 525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
