siRNA Details
Gene Symbol: NDUFA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193375.1 | NP_001180304.1 | 2 | 467 | ||
NM_175614.4 | NP_783313.1 | 2 | 467 | |||
NR_034166.2 | 3 | 587 | ||||
GenBank mRNA | AJ539081.1 | - | 2 | 162 | ||
AK127692.1 | - | 2 | 240 | |||
AL546175.3 | - | 3 | 328 | |||
AY726561.1 | - | 2 | 223 | |||
BC069045.1 | - | 2 | 222 | 6503302 | ||
BQ054257.1 | - | 2 | 223 | |||
BQ671956.1 | - | 2 | 241 | |||
BQ955016.1 | - | 2 | 213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
