siRNA Details
Gene Symbol: POTEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277304.1 | NP_001264233.1 | 1 | 415 | ||
NR_102391.1 | 1 | 415 | ||||
XM_006720355.3 | XP_006720418.1 | 1 | 529 | |||
XM_011543794.2 | XP_011542096.1 | 1 | 529 | |||
XM_017021858.1 | XP_016877347.1 | 1 | 529 |
Gene Symbol: LOC100288966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257362.1 | NP_001244291.1 | 1 | 545 | ||
XM_006709930.3 | XP_006709993.1 | 1 | 641 | |||
XM_011545655.2 | XP_011543957.1 | 1 | 641 | |||
XM_011545656.2 | XP_011543958.1 | 1 | 641 | |||
GenBank mRNA | AY172978.1 | - | 1 | 545 |
Gene Symbol: LOC102723502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724900.3 | XP_006724963.1 | 1 | 640 | ||
XM_006724901.3 | XP_006724964.1 | 1 | 640 | |||
XM_011546199.2 | XP_011544501.1 | 1 | 640 | |||
XM_011546200.2 | XP_011544502.1 | 1 | 640 |
Gene Symbol: POTEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277303.1 | NP_001264232.1 | 1 | 415 | ||
NR_102390.1 | 1 | 415 | ||||
XM_006720354.3 | XP_006720417.1 | 1 | 529 | |||
XM_011543793.2 | XP_011542095.1 | 1 | 529 | |||
XM_017021853.1 | XP_016877342.1 | 1 | 529 | |||
GenBank mRNA | BC101007.1 | - | 1 | 415 | 40006489 |
Gene Symbol: POTEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207355.3 | NP_997238.2 | 1 | 864 | ||
NR_110752.1 | 1 | 864 | ||||
XM_006720358.2 | XP_006720421.1 | 1 | 911 | |||
XM_011543795.2 | XP_011542097.1 | 1 | 911 | |||
XM_011543796.2 | XP_011542098.1 | 1 | 911 | |||
XM_011543797.2 | XP_011542099.1 | 1 | 911 | |||
GenBank mRNA | AY465170.1 | - | 2 | 434 | ||
AY465173.1 | - | 1 | 541 | |||
BC066981.1 | - | 1 | 864 | 4824630 | ||
BC101005.1 | - | 1 | 415 | 40006483 | ||
BC101006.1 | - | 1 | 526 | 40006485 | ||
BC127123.1 | - | 1 | 526 | 40006490 |
Gene Symbol: POTED | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174981.3 | NP_778146.2 | 1 | 545 | ||
XM_006723997.3 | XP_006724060.1 | 1 | 641 | |||
XM_011529550.2 | XP_011527852.1 | 1 | 641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
