siRNA Details
Gene Symbol: RASGEF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175062.3 | NP_778232.2 | 6 | 795 | ||
GenBank mRNA | AF449764.1 | - | 5 | 983 | ||
AK093160.1 | - | 3 | 647 | |||
BC036802.1 | - | 7 | 813 | |||
BC057759.1 | - | 3 | 449 | 5268021 |
