siRNA Details
Gene Symbol: EMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY761095.1 | - | 8 | 1018 | ||
BC032948.2 | - | 7 | 893 | 4794992 | ||
BC035001.2 | - | 7 | 891 | 4796476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
