siRNA Details
Gene Symbol: DPY19L2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003561.2 | 10 | 2449 | |||
NR_027768.1 | 7 | 2595 | ||||
GenBank mRNA | AK093485.1 | - | 2 | 76 | ||
AK131353.1 | - | 10 | 2442 | |||
AL834175.1 | - | 7 | 2348 | |||
BC150309.1 | - | 7 | 2056 |
Gene Symbol: DPY19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173812.4 | NP_776173.3 | 7 | 1009 | ||
XM_006719352.2 | XP_006719415.1 | 7 | 935 | |||
XM_011538215.2 | XP_011536517.1 | 5 | 475 | |||
XM_017019188.1 | XP_016874677.1 | 7 | 1056 | |||
XM_017019189.1 | XP_016874678.1 | 7 | 1056 | |||
XM_017019190.1 | XP_016874679.1 | 7 | 1056 | |||
XM_017019193.1 | XP_016874682.1 | 6 | 591 | |||
XM_017019194.1 | XP_016874683.1 | 6 | 618 | |||
XM_017019195.1 | XP_016874684.1 | 7 | 666 | |||
XM_017019196.1 | XP_016874685.1 | 7 | 1056 | |||
XM_017019197.1 | XP_016874686.1 | 6 | 889 | |||
XM_017019198.1 | XP_016874687.1 | 7 | 1056 | |||
XM_017019200.1 | XP_016874689.1 | 7 | 1056 | |||
XM_017019201.1 | XP_016874690.1 | 7 | 944 | |||
XM_017019202.1 | XP_016874691.1 | 6 | 557 | |||
XM_017019203.1 | XP_016874692.1 | 7 | 839 | |||
XM_017019204.1 | XP_016874693.1 | 8 | 1077 | |||
XM_017019205.1 | XP_016874694.1 | 5 | 931 | |||
GenBank mRNA | AK057511.1 | - | 7 | 980 | ||
AK310102.1 | - | 6 | 891 | |||
AL833344.1 | - | 7 | 936 | |||
AY358792.1 | - | 7 | 930 | |||
BC031225.1 | - | 11 | ||||
BC068442.1 | - | 7 | 1012 | 5297524 | ||
BC125215.1 | - | 7 | 899 | |||
BC125216.1 | - | 7 | 899 |
Gene Symbol: DPY19L2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002833.2 | 7 | 933 | |||
GenBank mRNA | AK304236.1 | - | 7 | 1093 | ||
BC066987.1 | - | 7 | 933 | |||
BX648666.1 | - | 2 | 100 |
Gene Symbol: DPY19L2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ287933.1 | - | 4 | 353 |
Gene Symbol: LOC107986726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012901.1 | XP_016868390.1 | 1 | 39 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
