siRNA Details
Gene Symbol: CES4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190201.1 | NP_001177130.1 | 8 | 992 | ||
NM_001190202.1 | NP_001177131.1 | 8 | 1004 | |||
NM_001318506.1 | NP_001305435.1 | 8 | 1048 | |||
NM_173815.6 | NP_776176.5 | 10 | 1311 | |||
XM_005255896.4 | XP_005255953.1 | 10 | 1308 | |||
XM_011523021.2 | XP_011521323.1 | 11 | 1440 | |||
XM_011523023.2 | XP_011521325.1 | 11 | 1428 | |||
XM_011523024.2 | XP_011521326.1 | 11 | 1440 | |||
XM_011523026.2 | XP_011521328.1 | 10 | 1320 | |||
XM_011523027.2 | XP_011521329.1 | 11 | 1440 | |||
XM_011523028.2 | XP_011521330.1 | 10 | 1328 | |||
XM_011523029.2 | XP_011521331.1 | 11 | 1440 | |||
XM_017023161.1 | XP_016878650.1 | 11 | 1428 | |||
XM_017023165.1 | XP_016878654.1 | 11 | 1428 | |||
XM_017023167.1 | XP_016878656.1 | 8 | 1392 | |||
GenBank mRNA | AK094783.1 | - | 8 | 1009 | ||
AK293061.1 | - | 8 | 840 | |||
AK295483.1 | - | 8 | 944 | |||
AK296064.1 | - | 10 | 1231 | |||
AK300792.1 | - | 8 | 952 | |||
BC064573.1 | - | 8 | 772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
