siRNA Details
Gene Symbol: TCAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282913.1 | NP_001269842.1 | 11 | 1534 | ||
NM_001282914.1 | NP_001269843.1 | 11 | 1640 | |||
NM_001282915.1 | NP_001269844.1 | 11 | 1458 | |||
NM_173826.3 | NP_776187.2 | 11 | 1716 | |||
XM_017006251.1 | XP_016861740.1 | 12 | 2668 | |||
GenBank mRNA | AK123680.1 | - | 2 | 113 | ||
AK124087.1 | - | 8 | 955 | |||
AK292756.1 | - | 11 | 1534 | |||
AL832473.1 | - | 11 | 1393 | |||
AL833118.1 | - | 10 | 1134 | |||
BC047231.1 | - | 11 | 1335 | 4817904 | ||
BC101058.2 | - | 11 | 2056 | |||
BC101059.2 | - | 11 | 1317 | 40008143 | ||
BC101060.2 | - | 11 | 1393 | 40008144 | ||
BC101061.2 | - | 11 | 1317 | 40008152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
